Order Entry
Norway
ContactUsLinkComponent
Anti-APBA3 Rabbit Polyclonal Antibody (Cy3®)
Anti-APBA3 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-11058R-CY3
Supplier:  Bioss
Anti-APBA3 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-11058R-CY3
Supplier:  Bioss

Specifications

  • Antistofftype:
    Primary
  • Antigen-navn:
    Amyloid Beta (A4) Precursor Protein-Binding, Family A, Member 3
  • Antigensymbol:
    APBA3
  • Klonalitet:
    Polyclonal
  • Konjugering:
    Cy3®
  • Vert:
    Rabbit
  • ImmunoKjemi:
    Yes
  • Isotype:
    IgG
  • Reaktivitet:
    Rat,
    Mouse
  • Kyssabsorpsjon:
    No
  • Skjema:
    liquid
  • Gen-ID:
    83611
  • Antigen synonymer:
    mint3|X11L2|MGC:15815
  • Lagringsbuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagringstemp:
    Store at 4°C for 12 months
  • Konsentrasjon:
    1 μg/μl
  • Frakttemp.:
    4°C
  • Immunogen:
    240-290/569
  • Rensing:
    Purified by Protein A
  • Fp:
    100 µl

Specifications

About this item

May modulate processing of the beta-amyloid precursor protein (APP) and hence formation of beta-APP. May enhance the activity of HIF1A in macrophages by inhibiting the activity of HIF1AN (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APBA3
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat