Anti-MFN1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-0557R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Mitofusin 1
- Antigen symbol:MFN1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:55669
- Antigen synonyms:hfzo1|hfzo2|MFN1|Fzo homolog|Mitofusin-1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:690-740/741
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Essential transmembrane GTPase, which mediates mitochondrial fusion. Fusion of mitochondria occurs in many cell types and constitutes an important step in mitochondria morphology, which is balanced between fusion and fission. MFN1 acts independently of the cytoskeleton. Overexpression induces the formation of mitochondrial networks.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MFN1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat