Anti-LDLR Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-0705R-CY3
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Low-Density Lipoprotein Receptor
- Antigen symbol:LDLR
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Multiple,Human,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:3949
- Antigen synonyms:FHC|FH|LDLCQ2|LDL receptor|LDL-R|Low-density lipoprotein receptor
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:811-850/860
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Binds LDL, the major cholesterol-carrying lipoprotein of plasma, and transports it into cells by endocytosis. In order to be internalized, the receptor-ligand complexes must first cluster into clathrin-coated pits. In case of HIV-1 infection, functions as a receptor for extracellular Tat in neurons, mediating its internalization in uninfected cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LDLR
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Other Species