Order Entry
Norway
ContactUsLinkComponent
Anti-ITGA5 Rabbit Polyclonal Antibody (Cy5.5®)
Anti-ITGA5 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-0567R-CY5.5
Supplier:  Bioss
Anti-ITGA5 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-0567R-CY5.5
Supplier:  Bioss

Specifications

  • Antistofftype:
    Primary
  • Antigen-navn:
    Integrin Alpha 5
  • Antigensymbol:
    ITGA5
  • Klonalitet:
    Polyclonal
  • Konjugering:
    Cy5.5®
  • Vert:
    Rabbit
  • ImmunoKjemi:
    Yes
  • Isotype:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Kyssabsorpsjon:
    No
  • Skjema:
    liquid
  • Gen-ID:
    3678
  • Antigen synonymer:
    FNRA|CD49e|Fibronectin Receptor Alpha Polypeptide|VLA-5|VLA5A
  • Lagringsbuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagringstemp:
    Store at 4°C for 12 months
  • Konsentrasjon:
    1 μg/μl
  • Frakttemp.:
    4°C
  • Immunogen:
    1000-1049/1049
  • Rensing:
    Purified by Protein A
  • Fp:
    100 µl

Specifications

About this item

Integrin alpha-5/beta-1 is a receptor for fibronectin and fibrinogen. It recognizes the sequence R-G-D in its ligands. In case of HIV-1 infection, the interaction with extracellular viral Tat protein seems to enhance angiogenesis in Kaposi's sarcoma lesions.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ITGA5
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat