Anti-TIMP3 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-0417R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Tissue Inhibitor of Metalloproteinases 3
- Antigen symbol:TIMP3
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:7078
- Antigen synonyms:K222TA2|tissue inhibitor of metalloproteinases 3|TIMP-3|MIG-5 protein|K222|HSMRK222|metalloproteinase inhibitor 3|protein MIG-5|SFD
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:150-200/211
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Complexes with metalloproteinases (such as collagenases) and irreversibly inactivates them by binding to their catalytic zinc cofactor. May form part of a tissue-specific acute response to remodeling stimuli. Known to act on MMP-1, MMP-2, MMP-3, MMP-7, MMP-9, MMP-13, MMP-14 and MMP-15.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TIMP3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat