Anti-GFAP Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-0199R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Glial fibrillary acidic Protein
- Antigen symbol:GFAP
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2670
- Antigen synonyms:FLJ45472|Intermediate filament protein.|Astrocyte|p-GFAP Ser38|GFAP phospho S38|GFAP|Glial Fibrillary Acidic Protein
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:30-80/432
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
GFAP, a class-III intermediate filament, is a cell-specific marker that, during the development of the central nervous system, distinguishes astrocytes from other glial cells.
Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200
Type: Primary
Antigen: GFAP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat