Order Entry
Norway
ContactUsLinkComponent
 
Anti-CK1 Chicken Polyclonal Antibody
Catalog # BSENC-1371-100
Supplier:  Biosensis
Anti-CK1 Chicken Polyclonal Antibody
Catalog # BSENC-1371-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Casein kinase 1 alpha
  • Antigen symbol:
    CK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Chicken
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Epitope:
    Shortest isoform of recombinant full length CK1a.
  • Format:
    Antibody is supplied as an aliquot of concentrated IgY preparation.
  • Form:
    Lyophilized
  • Gene ID:
    P48729 KC1A_HUMAN
  • Antigen synonyms:
    Casein kinase 1 alpha|CK1
  • Storage buffer:
    Contains 3% treahlose without preservatives.
  • Storage temperature:
    After reconstitution of lyophilised antibody, aliquot and store at −20 °C for a higher stability.
  • Shipping temperature:
    Room (ambient) temperature.
  • Immunogen:
    Shortest isoform of recombinant full length CK1a.
  • Purification:
    Antibody is supplied as an aliquot of concentrated IgY preparation.
  • Size:
    100 ul
  • Pk:
    100 µl

Specifications

About this item

Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. It can phosphorylate a large number of proteins. Participates in Wnt signaling. Phosphorylates CTNNB1 at 'Ser-45'. May phosphorylate PER1 and PER2. May play a role in segregating chromosomes during mitosis. May play a role in keratin cytoskeleton disassembly and thereby, it may regulate epithelial cell migration. (Reference: uniprot.org)

Western Blotting (WB): 1:5,000 - 1:10,000. Casein kinase 1 alpha has a predicted molecular weight of 38 kDa.

Immunocytochemistry (ICC): 1:500-1:1,000.

Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Shortest isoform of recombinant full length CK1a.
Host: Chicken
Isotype:
Reactivity: Human;Rat