Order Entry
Norway
ContactUsLinkComponent
Stopcock, three-way
Stopcock, three-way
Catalog # SCERSP845100-0004
Supplier:  DWK Life Sciences
undefined
Stopcock, three-way
Catalog # SCERSP845100-0004
Supplier:  DWK Life Sciences

Specifications

  • Hole Ø:
    4 mm
  • Pk:
    1
  • For tube O-Ø:
    10 mm

Specifications

About this item

Made of 33 expansion, low extractable borosilicate glass.

  • Very good chemical and temperature resistance
  • 1:10 taper
  • Hollow glass plug

Three-way, double-oblique bore, precision ground stopcock.Lower vacuum chamber is not in the inline of flow and must be outgassed in a separate operation.

Certifications: Confirms to USP Type I and ASTM E438, Type I, Class A requirements.