Human IgG Isotype Control
ICNA0855836
- Antibody type:Control
- Conjugation:Unconjugated
- Host:Human
- Isotype:IgG
- Form:Lyophilized from PBS, no azide
- Concentration:Total Protein: 10.5 mg/mL; IgG Concentration: 10.0 mg/mL. Packaged as 10 mg/mL
- Shipping temperature:Cold Pack
- Purification:Delipidation, ammonium sulfate fractionation and anion exchange chromatography. The resulting IgG fraction is dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, 0.05% sodium azide, adjusted to standard titer, filtered through a 0.22 µm filter, vialed and lyophilized.
- Size:1 ml
- Pk:1 mL
This product is the lyophilized powder of human IgG positive control and buffer salts. IgG is a purified antigen. Immunoglobulin G (IgG) is one of the most abundant proteins in serum. IgG is important for our defence against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions: to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.