Order Entry
Norway
ContactUsLinkComponent
Anti-C3 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-C3 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # ICNA0855237
Supplier:  MP Biomedicals
Anti-C3 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # ICNA0855237
Supplier:  MP Biomedicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Complement C3
  • Antigen symbol:
    C3
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Concentration:
    Total Protein: 28.8 mg/mL; Titer: 1:64,000
  • Shipping temperature:
    RT
  • Purification:
    Antibody and highly purified HRP (Rz>3.0) are conjugated under defined conditions to obtain optimally labeled product. Conjugated protein is purified by salt fractionation. The product is dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, adjusted to standard titer, filtered through a 0.22 µm filter, vialed and lyophilized.
  • Size:
    2 ml
  • Pk:
    2 mL

Specifications

About this item

Peroxidase-Conjugated Goat IgG Fraction to Human Complement C3 is the lyophilized powder of horseradish peroxidase (HRP)-conjugated goat IgG fraction to human complement C3 and buffer salts.

Type: Primary
Antigen: C3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human