Anti-MMP-8 Rabbit Polyclonal Antibody [clone: F445]
Catalog # BWRLBS1240
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:MMP-8
- Clonality:Polyclonal
- Clone:F445
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 415-460 of Human MMP-8.
MMP-8 (F445) pAb detects endogenous levels of MMP-8 protein.
Type: Primary
Antigen: MMP-8
Clonality: Polyclonal
Clone: F445
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat