Anti-MMP-2 Rabbit Polyclonal Antibody [clone: L638]
Catalog # BWRLBS1236
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:MMP-2
- Clonality:Polyclonal
- Clone:L638
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 601-650 of Human MMP-2.
MMP-2 (L638) pAb detects endogenous levels of MMP-2 protein.
Type: Primary
Antigen: MMP-2
Clonality: Polyclonal
Clone: L638
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat