Anti-MB67 Rabbit Polyclonal Antibody
Catalog # BWRLBS7155
Supplier: Bioworld Technology
Specifications
- Antistofftype:Primary
- Antigensymbol:MB67
- Klonalitet:Polyclonal
- Vert:Rabbit
- ImmunoKjemi:Yes
- Reaktivitet:Human
- Western blot:Yes
- Str:100 µg
- Fp:100 µG
Specifications
About this item
Recombinant full length Human MB67.
MB67 pAb detects endogenous levels of MB67 protein.
Type: Primary
Antigen: MB67
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human