Anti-p-Raf-1 Rabbit Polyclonal Antibody [clone: S259]
Catalog # BWRLBS4163
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-Raf-1
- Clonality:Polyclonal
- Clone:S259
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human Raf-1 around the phosphorylation site of Serine 259.
p-Raf1 (S259) pAb detects endogenous levels of Raf1 protein only when phosphorylated at Ser259
Type: Primary
Antigen: p-Raf-1
Clonality: Polyclonal
Clone: S259
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat