Order Entry
Norway
ContactUsLinkComponent
Anti-ATM Rabbit Polyclonal Antibody [clone: I1987]
Anti-ATM Rabbit Polyclonal Antibody [clone: I1987]
  BWRLBS5505
Anti-ATM Rabbit Polyclonal Antibody [clone: I1987]
  BWRLBS5505

 

  • Antibody type:
    Primary
  • Antigen symbol:
    ATM
  • Clonality:
    Polyclonal
  • Clone:
    I1987
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 1957-2006 of Human ATM.

ATM (I1987) pAb detects endogenous levels of ATM protein.

Type: Primary
Antigen: ATM
Clonality: Polyclonal
Clone: I1987
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human