Anti-p-NMDAzeta1 Rabbit Polyclonal Antibody [clone: S896]
Catalog # BWRLBS5090
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-NMDA zeta1
- Clonality:Polyclonal
- Clone:S896
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human NMDAzeta1 around the phosphorylation site of Serine 896.
p-NMDAR1 (S896) pAb detects endogenous levels of NMDAR1 protein only when phosphorylated at Ser896.
Type: Primary
Antigen: p-NMDAzeta1
Clonality: Polyclonal
Clone: S896
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat