Anti-p-IRS-1 Rabbit Polyclonal Antibody [clone: S612]
BWRLBS5084
- Antibody type:Primary
- Antigen symbol:p-IRS-1
- Clonality:Polyclonal
- Clone:S612
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human IRS-1 around the phosphorylation site of Serine 612.
p-IRS-1 (S612) pAb detects endogenous levels of IRS-1 protein only when phosphorylated at Ser612.
Type: Primary
Antigen: p-IRS-1
Clonality: Polyclonal
Clone: S612
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat