Anti-PDGFR-beta Rabbit Polyclonal Antibody [clone: D1003]
Catalog # BWRLBS1763
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PDGFR- beta
- Clonality:Polyclonal
- Clone:D1003
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 971-1020 of Human PDGFR-beta.
PDGFR-? (D1003) pAb detects endogenous levels of PDGFR-? protein.
Type: Primary
Antigen: PDGFR-beta
Clonality: Polyclonal
Clone: D1003
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat