Anti-IFN-R1 Rabbit Polyclonal Antibody [clone: L461]
Catalog # BWRLBS1754
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:IFN-R1
- Clonality:Polyclonal
- Clone:L461
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 410-460 of Human IFN-R1.
IFN-R1 (L461) pAb detects endogenous levels of Interferon alpha/beta receptor 1 protein.
Type: Primary
Antigen: IFN-R1
Clonality: Polyclonal
Clone: L461
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human