Anti-Histone H3 Rabbit Polyclonal Antibody [clone: A7]
Catalog # BWRLBS1750
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Histone H3
- Clonality:Polyclonal
- Clone:A7
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminal of Human Histone H3.
Histone H3 (A7) pAb detects endogenous levels of Histone H3 protein.
Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone: A7
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat