Anti-EphB1/2 Rabbit Polyclonal Antibody [clone: G590]
Catalog # BWRLBS1743
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:EphB1/2
- Clonality:Polyclonal
- Clone:G590
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 560-610 of Human EphB1.
EphB1/2 (G590) pAb detects endogenous levels of EphB1/2 protein.
Type: Primary
Antigen: EphB1/2
Clonality: Polyclonal
Clone: G590
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat