Anti-Histone H2B Rabbit Polyclonal Antibody [clone: P8]
Catalog # BWRLBS3153
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Histone H2B
- Clonality:Polyclonal
- Clone:P8
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids N-terminus of Human Histone H2B.
Histone H2B (P8) pAb detects endogenous levels of Histone H2B protein.
Type: Primary
Antigen: Histone H2B
Clonality: Polyclonal
Clone: P8
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse