Anti-GRB14 Rabbit Polyclonal Antibody [clone: D123]
Catalog # BWRLBS3130
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:GRB14
- Clonality:Polyclonal
- Clone:D123
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 90-102 of Human GRB14.
GRB14 (D123) pAb detects endogenous levels of GRB14 protein.
Type: Primary
Antigen: GRB14
Clonality: Polyclonal
Clone: D123
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat