Anti-INSL4 Rabbit Polyclonal Antibody [clone: L79]
Catalog # BWRLBS3118
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:INSL4
- Clonality:Polyclonal
- Clone:L79
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 34-96 of Human INSL4.
INSL4 (L79) pAb detects endogenous levels of INSL4 protein.
Type: Primary
Antigen: INSL4
Clonality: Polyclonal
Clone: L79
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human