Anti-Fos B Rabbit Polyclonal Antibody [clone: S23]
Catalog # BWRLBS1645
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Fos B
- Clonality:Polyclonal
- Clone:S23
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids N-terminus of Human Fos B.
FosB (S23) pAb detects endogenous levels of FosB protein.
Type: Primary
Antigen: Fos B
Clonality: Polyclonal
Clone: S23
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat