Anti-Catenin-delta1 Rabbit Polyclonal Antibody [clone: G224]
BWRLBS2992
- Antibody type:Primary
- Antigen symbol:Catenin- delta1
- Clonality:Polyclonal
- Clone:G224
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 201-250 of Human Catenin-delta1.
Catenin-?1 (G224) pAb detects endogenous levels of Catenin-?1 protein.
Type: Primary
Antigen: Catenin-delta1
Clonality: Polyclonal
Clone: G224
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat