Anti-PDE4B/C/D Rabbit Polyclonal Antibody [clone: H47]
BWRLBS1572
- Antibody type:Primary
- Antigen symbol:PDE4B/C/D
- Clonality:Polyclonal
- Clone:H47
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to the N-terminus of Human PDE4B/C/D.
PDE4B/C/D (H47) pAb detects endogenous levels of PDE4B/C/D protein.
Type: Primary
Antigen: PDE4B/C/D
Clonality: Polyclonal
Clone: H47
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat