Anti-Synapsin I Rabbit Polyclonal Antibody [clone: Y3]
BWRLBS3667
- Antibody type:Primary
- Antigen symbol:Synapsin I
- Clonality:Polyclonal
- Clone:Y3
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to the N-terminus of Human Synapsin I.
Synapsin I (Y3) pAb detects endogenous levels of Synapsin I protein.
Type: Primary
Antigen: Synapsin I
Clonality: Polyclonal
Clone: Y3
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat