Anti-Adducin gamma Rabbit Polyclonal Antibody [clone: P452]
Catalog # BWRLBS2209
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Adducin gamma
- Clonality:Polyclonal
- Clone:P452
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 420-471 of Human Adducin gamma.
Adducin ? (P452) pAb detects endogenous levels of Adducin ? protein.
Type: Primary
Antigen: Adducin gamma
Clonality: Polyclonal
Clone: P452
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat