Anti-HEN1/2 Rabbit Polyclonal Antibody [clone: A79]
BWRLBS2031
- Antibody type:Primary
- Antigen symbol:HEN1/2
- Clonality:Polyclonal
- Clone:A79
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 50-100 of Human HEN1.
HEN1/2 (A79) pAb detects endogenous levels of HEN1/2 protein.
Type: Primary
Antigen: HEN1/2
Clonality: Polyclonal
Clone: A79
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse