Anti-MRP-S36 Rabbit Polyclonal Antibody [clone: V36]
Catalog # BWRLBS3881
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:MRP-S36
- Clonality:Polyclonal
- Clone:V36
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminal of Human MRP-S36.
MRP-S36 (V36) pAb detects endogenous levels of MRP-S36 protein.
Type: Primary
Antigen: MRP-S36
Clonality: Polyclonal
Clone: V36
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human