Anti-VEGFR2/Flk-1 Rabbit Polyclonal Antibody [clone: D1209]
BWRLBS1372
- Antibody type:Primary
- Antigen symbol:VEGFR2/Flk-1
- Clonality:Polyclonal
- Clone:D1209
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 1209-1250 of Human VEGFR2.
VEGFR2 (D1209) pAb detects endogenous levels of VEGFR2 protein.
Type: Primary
Antigen: VEGFR2/Flk-1
Clonality: Polyclonal
Clone: D1209
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat