Anti-Synaptotagmin Rabbit Polyclonal Antibody [clone: M303]
Catalog # BWRLBS1347
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Synaptotagmin
- Clonality:Polyclonal
- Clone:M303
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 280-330 of Human Synaptotagmin.
Synaptotagmin (M303) pAb detects endogenous levels of Synaptotagmin protein.
Type: Primary
Antigen: Synaptotagmin
Clonality: Polyclonal
Clone: M303
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat