Anti-Synaptophysin Rabbit Polyclonal Antibody [clone: L128]
Catalog # BWRLBS1345
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Synaptophysin
- Clonality:Polyclonal
- Clone:L128
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 101-150 of Human Synaptophysin.
Synaptophysin (L128) pAb detects endogenous levels of Synaptophysin protein.
Type: Primary
Antigen: Synaptophysin
Clonality: Polyclonal
Clone: L128
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat