Anti-Stat6 Rabbit Polyclonal Antibody [clone: R639]
Catalog # BWRLBS1341
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Stat6
- Clonality:Polyclonal
- Clone:R639
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 610-660 of Human Stat6.
STAT6 (R639) pAb detects endogenous levels of STAT6 protein.
Type: Primary
Antigen: Stat6
Clonality: Polyclonal
Clone: R639
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat