Anti-APC Rabbit Polyclonal Antibody [clone: Q2829]
Catalog # BWRLBS1017
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:APC
- Clonality:Polyclonal
- Clone:Q2829
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the C-terminal of Human APC.
APC (Q2829) pAb detects endogenous levels of APC protein.
Type: Primary
Antigen: APC
Clonality: Polyclonal
Clone: Q2829
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat