Order Entry
Norway
ContactUsLinkComponent
 
Anti-PML Rabbit Polyclonal Antibody [clone: P39]
Catalog # BWRLBS1296
Anti-PML Rabbit Polyclonal Antibody [clone: P39]
Catalog # BWRLBS1296

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    PML
  • Clonality:
    Polyclonal
  • Clone:
    P39
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to the N-terminal of Human PML.

PML (P39) pAb detects endogenous levels of PML protein.

Type: Primary
Antigen: PML
Clonality: Polyclonal
Clone: P39
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human