Order Entry
Norway
ContactUsLinkComponent
Test strips, fluoride
Test strips, fluoride
Catalog # MANA90734
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test strips, fluoride
Catalog # MANA90734
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Alizarin red
  • Pk:
    30
  • Description:
    Fluoride test paper
  • Tests per kit:
    30
  • Range:
    0 · 2 · 5 · 10 · 20 · 50 · 100 mg/l F-

Specifications

About this item

These test discs and reagent are for the semi-quantitative determination of fluoride ions. The fluoride test is especially suited for rapid control of fluoride concentrations. Its purpose is to define concentration ranges and to detect values which fall short of or exceed given limits.

  • Colour change from red to white

Chlorates and bromates cause white discolourations of the test paper. Addition of sodium dithionite (Na₂S₂O₄) will circumvent this interference without affecting the fluoride test.

Packaging: Box of 30 test discs with reagent.