Order Entry
Norway
ContactUsLinkComponent
Accessories
Accessories
Catalog # ORIO1010005
Supplier:  Thermo Orion
Accessories
Catalog # ORIO1010005
Supplier:  Thermo Orion
Specifications
  • Description:
    Cable adapter, for connecting the RS232 port of a meter to a USB port
  • Seq:
    108
  • Pk:
    1
  • For:
    Orion™ Versa Star, Orion™ Star A series (A320, A220 + A210 series) and Orion™ Star and Star Plus meters
Specifications
About this item
This cable adapter can be used to connect a USB port to the RS232 port of a meter.
The adapter allows interface between a USB connection and Thermo Scientific™ Orion™ Versa Star meters, Thermo Scientific™ Orion™ Star A series (Star A320, Star A220 and Star A210 series) meters and legacy Orion™ Star and Star Plus meters.