Capillary columns, SolGel-1ms™
Supplier: Trajan Scientific and Medical
Certificates
About this item
A robust, high temperature column for mass spectrometers. Can also be used for all non MS detectors.
100% dimethyl polysiloxane.
Maximum temperature: 340/360 °C
Suitable replacement for: DB-1, DB-Petro, HP-1, HP-1MS, Rtx-1, Ultra-1, SPB-1, SPB-1 Sulfur, Petrocol DH, CP-Sil 5C, CP-Sil 5CB, VB-1, ZB-1.