Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Ordreinnføring
Ordrestatus
Din konto
Handlevogn
Norway
Ordreinnføring
Ordrestatus
Din konto
Handlevogn
ContactUsLinkComponent
Home
Glycine
Glycine ≥99.5%
Glycine ≥99.5%
:
MP Biomedicals
Glycine ≥99.5%
:
MP Biomedicals
:
56-40-6
:
:
NH₂CH₂COOH
:
MFCD00008131
:
75.07 g/mol
:
Ambient