Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Iodo-2'-deoxyuridine (Idoxuridine)
5-Iodo-2'-deoxyuridine (Idoxuridine) ≥99%, crystalline
5-Iodo-2'-deoxyuridine (Idoxuridine) ≥99%, crystalline
5-Iodo-2'-deoxyuridine (Idoxuridine) ≥99%, crystalline
Supplier:
MP Biomedicals
Certificates
Expand image
5-Iodo-2'-deoxyuridine (Idoxuridine) ≥99%, crystalline
Supplier:
MP Biomedicals
About this item
CAS Number:
54-42-2
:
Formula:
C₉H₁₁IN₂O₅
MDL Number:
MFCD00134656
:
354.10 g/mol
:
GHS07
:
GHS08