3,3'-Diaminobenzidine (DAB)
Catalog # ICNA08980681
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:100
- Seq:0100
Specifications
About this item
:
:214.27 g/mol
:GHS03
:GHS05
:GHS08
CAS Number:91-95-2
Formula:C₁₂H₁₄N₄
MDL Number:MFCD00007725