Order Entry
Norway
ContactUsLinkComponent
Aluminium oxide, Ecochrom™ Alumina N Activity: Super I
Aluminium oxide, Ecochrom™ Alumina N Activity: Super I
Catalog # ICNA0204589
Supplier:  MP Biomedicals
CAS Number:  
Aluminium oxide, Ecochrom™ Alumina N Activity: Super I
Catalog # ICNA0204589
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    50 kg
  • Seq:
    4418

Specifications

About this item

 :
 :101.96 g/mol
Density:3.94 g/cm³ (20 °C) (lit.)
Storage Temperature:Ambient
CAS Number:1344-28-1
 :11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424