Aluminium oxide, Ecochrom™ Alumina N Activity: Super I
Catalog # ICNA0204586
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:5 kg
- Seq:4114
Specifications
About this item
:
:101.96 g/mol
Density:3.94 g/cm³ (20 °C)
Storage Temperature:Ambient
CAS Number:1344-28-1
:11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424