Aluminiumoksid, Ecochrom™
Catalog # ICNA0202029
Supplier: MP Biomedicals
CAS Number:
Specifications
- Fp:50 kg
- Seq:4418
Specifications
About this item
:
:101.96 g/mol
Density:3,94 g/cm³ (20 °C) (lit.)
Storage Temperature:Romtemperatur
CAS Number:1344-28-1
:11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424