Order Entry
Norway
ContactUsLinkComponent
Dog plasma
Dog plasma
Catalog # S2930-100
Supplier:  Biowest
CAS Number:  
Dog plasma
Catalog # S2930-100
Supplier:  Biowest
CAS Number:  
Specifications
  • Pk:
    100 µl
  • Description:
    Dog (canine) plasma with sodium heparin
Specifications
About this item
Plasma provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth. It is used as a component of bioassays, immunoassays or enzyme assays.