CL-XPosure™ Film
Supplier: Thermo Fisher Scientific
Certificates
About this item
CL-XPosure™ Film is a clear blue X-ray film recommended for the detection and documentation of blotting assays.
- High performance - just as sensitive as other commercially available autoradiography films
- Economical - achieve high-quality images at an affordable price
- Convenient - available in five sheet sizes in packages containing 100 non-interleaved sheets
- Perfect match - validated for use with SuperSignal chemiluminescent substrates and Pierce ECL and ECL Plus chemiluminescent substrates
CL-XPosure™ works with chemiluminescent horseradish peroxidase (HRP) or alkaline phosphatase (AP) detection reagents as well as radioactivity, making it ideal for Western, Southern and Northern blots and Electrophoretic Mobility Shift Assays (EMSA). CL-XPosure™ Film exhibits the same detection sensitivity as other commercially available film brands.
CL-XPosure film is an excellent photographic film for use with enhanced chemiluminescence (ECL) substrates for horseradish peroxidase (HRP) or alkaline phosphatase (AP).
Delivery: Supplied in packs of 25, 50 or 100 non-interleaved sheets.