Folding tray, PVC
Catalog # CEMG10183
Supplier: CEMO
Specifications
- Capacity:30 l
- W×D×H:600×400×250 mm
- Pk:1
Specifications
About this item
This folding collection tray with built-in reinforcement plates and its extremely flat design helps to be transported in any vehicle. It is ideally suited for transporting hazardous materials on construction, agricultural and forestry machines.
- Low space requirement when unfolded (only 25 mm height)
- Quick assembly with Velcro fasteners
- Absolutely impervious to liquid
- Resistant to many chemicals and oils