Agarose, immunodiffusion grade, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Agarose is the most popular medium for immunoelectrophoresis because of the large pore size for rapid diffusion, and for low background staining by Coomassie Blue G stain. The low EEO and high gel strength is specifically selected and tested for immunoelectrophoresis and immunodiffusion.